Learn More
Medchemexpress LLC Diphenyl phthalate-3,4,5,6-d4 | 1398065-61-6 | MFCD22565776 | >98% | 5 MG
SDP
Catalog No. 5000409999
Encompass_Preferred
Diphenyl phthalate-3,4,5,6-d4 is the deuterium labeled Diphenyl phthalate. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, or LC-MS applications.
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-09999 Supplier Medchemexpress LLC Supplier No. HYB1966S5MG
Diphenyl phthalate-3,4,5,6-d4 is the deuterium labeled Diphenyl phthalate. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuterium substitution has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Serves as an internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, or LC-MS applications.
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles of drugs.