Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl Sulfoxide, MP Biomedicals

Catalog No. ICN208314 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN208314 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN208314 Supplier MP Biomedicals, Inc Supplier No. 0520831410

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 945-51-7
Molecular Formula C12H10OS
Molecular Weight (g/mol) 202.271
InChI Key JJHHIJFTHRNPIK-UHFFFAOYSA-N
Synonym diphenyl sulfoxide, phenyl sulfoxide, sulfinyldibenzene, phenylsulfinylbenzene, diphenyl sulphoxide, sulfoxide, diphenyl, diphenylsulfoxide, benzene, 1,1'-sulfinylbis, 1,1'-sulfinyldibenzene, phenylsulfinyl benzene
PubChem CID 13679
IUPAC Name benzenesulfinylbenzene
SMILES C1=CC=C(C=C1)S(=O)C2=CC=CC=C2

Specifications

Quantity 10 g
Formula Weight 202.28
Chemical Name or Material Diphenyl Sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.