Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl Sulfoxide 99.0+%, TCI America™
SDP

Catalog No. D100225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D100225G 25 g
D10025G 5 g
D1002100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D100225G Supplier TCI America Supplier No. D100225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 945-51-7
Molecular Formula C12H10OS
Molecular Weight (g/mol) 202.271
MDL Number MFCD00002085
InChI Key JJHHIJFTHRNPIK-UHFFFAOYSA-N
Synonym diphenyl sulfoxide, phenyl sulfoxide, sulfinyldibenzene, phenylsulfinylbenzene, diphenyl sulphoxide, sulfoxide, diphenyl, diphenylsulfoxide, benzene, 1,1'-sulfinylbis, 1,1'-sulfinyldibenzene, phenylsulfinyl benzene
PubChem CID 13679
IUPAC Name benzenesulfinylbenzene
SMILES C1=CC=C(C=C1)S(=O)C2=CC=CC=C2

Specifications

Melting Point 71°C
Color White
Boiling Point 198°C
Quantity 25 g
Formula Weight 202.27
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Sulfoxide
RTECSNumber DA9185000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.