Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylacetaldehyde 95.0+%, TCI America™
SDP

Catalog No. D24925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24925G 5 g
D249225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24925G Supplier TCI America Supplier No. D24925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 947-91-1
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD00006972
InChI Key HLLGFGBLKOIZOM-UHFFFAOYSA-N
Synonym diphenylacetaldehyde, diphenylketen, acetaldehyde, diphenyl, alpha-phenylbenzeneacetaldehyde, diphenylethanal, diphenyl-acetaldehyde, benzeneacetaldehyde, .alpha.-phenyl, diphenyl-acetaldehyd, benzeneacetaldehyde, ?-phenyl, diphenylacetoaldehyde
PubChem CID 13696
IUPAC Name 2,2-diphenylacetaldehyde
SMILES O=CC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Boiling Point 175°C
Quantity 5 g
Formula Weight 196.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diphenylacetaldehyde
RTECSNumber AB2827500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.