Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylacetic Anhydride 95.0+%, TCI America™
SDP

Catalog No. D40755G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40755G Supplier TCI America Supplier No. D40755G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1760-46-9
Molecular Formula C28H22O3
Molecular Weight (g/mol) 406.481
InChI Key YZMRCMTTYLBDPD-UHFFFAOYSA-N
PubChem CID 137197
IUPAC Name (2,2-diphenylacetyl) 2,2-diphenylacetate
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)OC(=O)C(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 98°C
Color White
Boiling Point 182°C
Quantity 5 g
Formula Weight 406.48
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylacetic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.