Learn More
Cayman Chemical DIphenylamIn 100mg
SDP
Catalog No. 502936391
Encompass_Preferred
An antioxidant and a pesticide; inhibits the oxidation of α-farnesene in solution as well as in the coating of Granny Smith apples postharvest; prevents superficial scald in several varieties of apples when applied by dipping or wrapping postharvest at 1,000 ppm; active against various benomyl-resistant or -sensitive isolates of the plant pathogenic fungus P. expansum (EC50s = <2-12.8 and 22.9-78.5 UG/ml, respectively); induces mortality in sheep keds at 0.25%; has been found as a contaminant in various jellies and jams and seawater
Catalog No. 50-293-6391 Supplier Cayman Chemical Supplier No. 24250100 MG
An antioxidant and a pesticide; inhibits the oxidation of α-farnesene in solution as well as in the coating of Granny Smith apples postharvest; prevents superficial scald in several varieties of apples when applied by dipping or wrapping postharvest at 1,000 ppm; active against various benomyl-resistant or -sensitive isolates of the plant pathogenic fungus P. expansum (EC50s = <2-12.8 and 22.9-78.5 UG/ml, respectively); induces mortality in sheep keds at 0.25%; has been found as a contaminant in various jellies and jams and seawater