Learn More
Selleck Chemical LLC Diphenylamine Hydrochloride
SDP
Catalog No. 5000003811
Encompass_Preferred
Diphenylamine (N-Phenylbenzenamine Anilinobenzene C I 10355) a derivative of aniline is widely used as an industrial antioxidant dye mordant and reagent and is also employed in agriculture as a fungicide and antihelmintic Diphenylamine can inhibits oxidative phosphorylationsignificantly although its inhibition potency was weaker than that of NSAIDs with a diphenylamine structure
Catalog No. 50-000-03811 Supplier Selleck Chemical LLC Supplier No. S560425MG
Diphenylamine (N-Phenylbenzenamine Anilinobenzene C I 10355) a derivative of aniline is widely used as an industrial antioxidant dye mordant and reagent and is also employed in agriculture as a fungicide and antihelmintic Diphenylamine can inhibits oxidative phosphorylationsignificantly although its inhibition potency was weaker than that of NSAIDs with a diphenylamine structure