Learn More
Medchemexpress LLC Diphenylamine hydrochloride | 537-67-7 | 1 G
SDP
Catalog No. 5000418130
Encompass_Preferred
Diphenylamine hydrochloride (N-Phenylaniline hydrochloride) is an antihyperglycemic agent with oral activity and shares a common structure with non-steroidal anti-inflammatory drugs (NSAIDs). It functions by uncoupling oxidative phosphorylation in mitochondria, which can lead to a decrease in hepatic cell ATP levels and potential liver cell damage. Additionally, it serves as an industrial antioxidant, a dyeing mordant, and finds use in agriculture as both an antifungal and antibacterial agent.
  • Antihyperglycemic agent with oral activity
  • Functions as an industrial antioxidant
  • Acts as a dyeing mordant
  • Used as an antifungal agent in agriculture
  • Used as an antibacterial agent in agriculture
  • Uncouples oxidative phosphorylation in mitochondria
  • Leads to decreased hepatic cell ATP levels
Catalog No. 50-004-18130 Supplier Medchemexpress LLC Supplier No. HYN71331G
Diphenylamine hydrochloride (N-Phenylaniline hydrochloride) is an antihyperglycemic agent with oral activity and shares a common structure with non-steroidal anti-inflammatory drugs (NSAIDs). It functions by uncoupling oxidative phosphorylation in mitochondria, which can lead to a decrease in hepatic cell ATP levels and potential liver cell damage. Additionally, it serves as an industrial antioxidant, a dyeing mordant, and finds use in agriculture as both an antifungal and antibacterial agent.
  • Antihyperglycemic agent with oral activity
  • Functions as an industrial antioxidant
  • Acts as a dyeing mordant
  • Used as an antifungal agent in agriculture
  • Used as an antibacterial agent in agriculture
  • Uncouples oxidative phosphorylation in mitochondria
  • Leads to decreased hepatic cell ATP levels