Learn More
Medchemexpress LLC Diphenylamine hydrochloride | 537-67-7 | 1 ML
SDP
Catalog No. 5000417594
Encompass_Preferred
Diphenylamine hydrochloride (N-Phenylaniline hydrochloride) is an antihyperglycemic agent with oral activity. It functions as an industrial antioxidant, a dyeing mordant, and an antifungal and antibacterial agent in agriculture. It is intended for research use only and not sold to patients.
  • Shares a common structure with non-steroidal anti-inflammatory drugs (NSAIDs)
  • Works by uncoupling oxidative phosphorylation in mitochondria
  • Can lead to a decrease in hepatic cell ATP levels and may cause liver cell damage
  • Excreted through urine and bile in rats
  • Reduced blood sugar by 61.4% in hyperglycemic rabbits at 10 mg/kg oral dose
Catalog No. 50-004-17594 Supplier Medchemexpress LLC Supplier No. HYN713310MM/1ML
Diphenylamine hydrochloride (N-Phenylaniline hydrochloride) is an antihyperglycemic agent with oral activity. It functions as an industrial antioxidant, a dyeing mordant, and an antifungal and antibacterial agent in agriculture. It is intended for research use only and not sold to patients.
  • Shares a common structure with non-steroidal anti-inflammatory drugs (NSAIDs)
  • Works by uncoupling oxidative phosphorylation in mitochondria
  • Can lead to a decrease in hepatic cell ATP levels and may cause liver cell damage
  • Excreted through urine and bile in rats
  • Reduced blood sugar by 61.4% in hyperglycemic rabbits at 10 mg/kg oral dose