Learn More
Hach Company Diphenylcarbazone Reagent, 113 g
Supplier:  Hach Company 83514
Encompass
Mixed indicator-buffer for chloride determination by Hach mercuric nitrate titration methods. Bulk Powder.
Catalog No. 50-199-4454