Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylcarbazone Indicator, 0.1% (w/v) Alcoholic Solution, Ricca Chemical
SDP

Catalog No. 26004 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2600-4 120 mL Amber Glass Bottle
2600-16 500 mL Amber Glass Bottle
R26000001C 1 L Amber Glass Bottle
R26000004C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 2600-4 Supplier Ricca Chemical Company Supplier No. 26004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Diphenylcarbazone Indicator
CAS 64-17-5, 67-56-1, 538-62-5, 140-22-7
CAS Min % 0.12, 98.0
CAS Max % 0.13, 100.0
Concentration 0.1% (w/v)
Packaging Amber Glass Bottle
Molecular Formula C13H12N4O
Solvent or Matrix Alcoholic Solution
Synonym 1,5-diphenylcarbazone, diazenecarboxylic acid, phenyl-, 2-phenylhydrazide, unii-5uyj4r0d5t, phenylazoformic acid 2-phenylhydrazide, sym-diphenylcarbazone, 5uyj4r0d5t, chembl79409, 3-hydroxy-1,5-diphenylformazan, phenyldiazenecarboxylic acid 2-phenylhydrazide, 3-phenylamino-1-phenylimino urea
Color Yellow
Quantity 120 mL
InChI Key ZFWAHZCOKGWUIT-UHFFFAOYSA-N
Physical Form Liquid
SMILES O=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
IUPAC Name 3-(phenylamino)-1-(phenylimino)urea
Molecular Weight (g/mol) Mixture
PubChem CID 10860
Percent Purity 0.1% W/V
Grade Indicator
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.