Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylcarbazone Indicator, 0.1% (w/v) Alcoholic Solution, Ricca Chemical
SDP

Catalog No. R26000004C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R26000004C 4 L Amber Glass Bottle
2600-4 120 mL Amber Glass Bottle
2600-16 500 mL Amber Glass Bottle
R26000001C 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R26000004C Supplier Ricca Chemical Company Supplier No. 26001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Diphenylcarbazone Indicator
CAS 64-17-5, 67-56-1, 538-62-5, 140-22-7
CAS Min % 98.0, 0.12
CAS Max % 100.0, 0.13
Concentration 0.1% (w/v)
Packaging Amber Glass Bottle
Solvent or Matrix Alcoholic Solution
Color Yellow
Quantity 4 L
Physical Form Liquid
Percent Purity 0.1% W/V
Grade Indicator
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.