Learn More
Hach Company Diphenylcarbazone Reagent Powder Pillows, pk/100
Supplier:  Hach Company 96799
Encompass
Indicator-buffer reagent for determination of chloride by Hach mercuric nitrate titration methods. Pack of 100 Powder Pillows.
Catalog No. 50-199-4478