Learn More
Hach Company Diphenylcarbazone Reagent Powder Pillows, 100mL sample, pk/100
Catalog No. 501994455 Shop All Hach Company Products
Encompass
Indicator-buffer reagent for chloride determination by Hach mercuric nitrate titration methods. Hach method 8206. Powder pillows for 100-mL sample. Pack of 100 powder pillows.
Catalog No. 50-199-4455 Supplier Hach Company Supplier No. 83699
Indicator-buffer reagent for chloride determination by Hach mercuric nitrate titration methods. Hach method 8206. Powder pillows for 100-mL sample. Pack of 100 powder pillows.