Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylcarbazone-Xylene Cyanol FF, For Chloride, Certified, LabChem™

Catalog No. LC136827 Shop All LabChem Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC136827 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC136827 Supplier LabChem, Inc. Supplier No. LC136827
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Diphenylcarbazone-Xylene Cyanol FF
Color Blue
Name Note For Chloride
CAS Max % 99.72, 0.25, 0.03
CAS 538-62-5, 140-22-7, 4463-44-9
Linear Formula C13H14N4O
UN Number UN1993
Solubility Information Soluble in water
Formula Weight 242.28
Grade Certified
Identification Passes Test
Packaging Amber Glass
Suitability Passes Test
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; May release flammable gases; Sulfur compounds
Quantity 125 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.