Learn More
Medchemexpress LLC HY-100965 10mg , Diphenyleneiodonium chloride CAS:4673-26-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10096510MG
Encompass_Preferred
HY-100965 10mg Diphenyleneiodonium chloride CAS: 4673-26-1 Diphenyleneiodonium chloride is a NADPH oxidase (NOX) inhibitor and also functions as a TRPA1 activator with an EC50 of 1 to 3 μM. Diphenyleneiodonium chloride selectively inhibits intracellular reactive oxygen species. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9594