Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyliodonium-2-carboxylate Monohydrate 98.0+%, TCI America™
SDP

Catalog No. D25035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25035G 5 g
D250325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25035G Supplier TCI America Supplier No. D25035G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96195-89-0
Molecular Formula C13H11IO3
Molecular Weight (g/mol) 342.132
MDL Number MFCD00150328
InChI Key NHCFFVMWVHFFSJ-UHFFFAOYSA-N
Synonym (2-Carboxyphenyl)phenyliodonium Hydroxide Inner Salt
PubChem CID 22271963
IUPAC Name 2-phenyliodoniobenzoate;hydrate
SMILES C1=CC=C(C=C1)[I+]C2=CC=CC=C2C(=O)[O-].O

Specifications

Color White
Quantity 5 g
Formula Weight 324.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyliodonium-2-carboxylate Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.