Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyliodonium hexafluorophosphate, 98%, Thermo Scientific™

Catalog No. AA1890606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1890606 5 g
AA1890614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1890606 Supplier Thermo Scientific Chemicals Supplier No. 1890606

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58109-40-3
Molecular Formula C12H10I
Molecular Weight (g/mol) 281.12
MDL Number MFCD00061398
InChI Key OZLBDYMWFAHSOQ-UHFFFAOYSA-N
Synonym diphenyliodonium hexafluorophosphate, diphenyliodanium hexafluorophosphate, diphenyliodonium hexafluorophosphate v, iodonium, diphenyl-, hexafluorophosphate 1-, iodonium, diphenyl-, hexafluorophosphate 1-1:1, diphenyliodonium pf6, dpihfp, acmc-209m2o, phi + pf6-, diphenyliodanium; hexafluoro-$l^ 5-phosphanuide
PubChem CID 2737136
IUPAC Name diphenyliodanium
SMILES [I+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 138°C to 140°C
Quantity 5 g
UN Number UN2811
Formula Weight 426.08
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Diphenyliodonium hexafluorophosphate
Hazard Category Danger
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.