Learn More
Medchemexpress LLC Diphenyliodonium iodide | 2217-79-0 | MFCD00011910 | 98.0% | 408.02 | C12H10I2 | 10 G
SDP
Catalog No. 5000281397
Encompass_Preferred
Diphenyliodonium iodide is a research-grade chemical reagent that functions as an inhibitor of reactive oxygen species (ROS). It is used in biochemical assays and preservation studies to probe ROS-dependent pathways and effects on phenylpropanoid metabolism. The material is supplied as a solid with recommended cold, sealed storage to maintain stability.
  • Inhibits reactive oxygen species (ROS) in biochemical assays.
  • High purity suitable for research (98.0%).
  • Solid form; store sealed at low temperature to preserve stability.
  • Applicable to studies of phenylpropanoid pathway and preservation effects.
  • Available in multiple laboratory pack sizes for experimental needs.
Catalog No. 50-002-81397 Supplier Medchemexpress LLC Supplier No. HYW25030710G
Diphenyliodonium iodide is a research-grade chemical reagent that functions as an inhibitor of reactive oxygen species (ROS). It is used in biochemical assays and preservation studies to probe ROS-dependent pathways and effects on phenylpropanoid metabolism. The material is supplied as a solid with recommended cold, sealed storage to maintain stability.
  • Inhibits reactive oxygen species (ROS) in biochemical assays.
  • High purity suitable for research (98.0%).
  • Solid form; store sealed at low temperature to preserve stability.
  • Applicable to studies of phenylpropanoid pathway and preservation effects.
  • Available in multiple laboratory pack sizes for experimental needs.