Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diphenyliodonium iodide | 2217-79-0 | MFCD00011910 | 98.0% | 408.02 g/mol | C12H10I2 | 1 G
SDP

Catalog No. 5000281819
Encompass_Preferred

Diphenyliodonium iodide is an organoiodine research reagent used primarily as an inhibitor of reactive oxygen species (ROS) in biochemical studies and as a preservative in select applications. Supplied as a high-purity solid, it is intended for laboratory research use and requires moisture-controlled storage.

  • Inhibits reactive oxygen species for oxidative stress studies.
  • High purity (98.0%) suitable for research applications.
  • Molecular formula C12H10I2 and molecular weight 408.02 g/mol.
  • Solid, white to yellow appearance for easy handling.
  • Storage at 4°C, sealed and dry; solvent storage at -80°C or -20°C for limited durations.
  • Available as solid quantities and DMSO solutions for flexible use.

Catalog No. 50-002-81819 Supplier Medchemexpress LLC Supplier No. HYW2503071G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diphenyliodonium iodide is an organoiodine research reagent used primarily as an inhibitor of reactive oxygen species (ROS) in biochemical studies and as a preservative in select applications. Supplied as a high-purity solid, it is intended for laboratory research use and requires moisture-controlled storage.

  • Inhibits reactive oxygen species for oxidative stress studies.
  • High purity (98.0%) suitable for research applications.
  • Molecular formula C12H10I2 and molecular weight 408.02 g/mol.
  • Solid, white to yellow appearance for easy handling.
  • Storage at 4°C, sealed and dry; solvent storage at -80°C or -20°C for limited durations.
  • Available as solid quantities and DMSO solutions for flexible use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.