Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diphenyliodonium iodide | 2217-79-0 | MFCD00011910 | ≥98.0% | 408.02 g·mol⁻¹ | C12H10I2 | 1 ML
SDP

Catalog No. 5000281239
Encompass_Preferred

Diphenyliodonium iodide is a research reagent that acts as an inhibitor of reactive oxygen species (ROS). It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and is intended for laboratory research use only.

  • Inhibitor of reactive oxygen species (ROS).
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • High purity (≥98.0%).
  • Applicable to studies of oxidative stress and preservation mechanisms.
  • Chemical formula C12H10I2; molecular weight 408.02 g·mol⁻¹; CAS 2217-79-0.

Catalog No. 50-002-81239 Supplier Medchemexpress LLC Supplier No. HYW25030710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diphenyliodonium iodide is a research reagent that acts as an inhibitor of reactive oxygen species (ROS). It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and is intended for laboratory research use only.

  • Inhibitor of reactive oxygen species (ROS).
  • Supplied as a 10 mM solution in DMSO, 1 mL.
  • High purity (≥98.0%).
  • Applicable to studies of oxidative stress and preservation mechanisms.
  • Chemical formula C12H10I2; molecular weight 408.02 g·mol⁻¹; CAS 2217-79-0.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.