Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Diphenyliodonium iodide | 2217-79-0 | MFCD00011910 | 98.0% | 408.02 g/mol | C12H10I2 | 25 G
SDP

Catalog No. 5000281229
Encompass_Preferred

Diphenyliodonium iodide is a research-grade reagent used primarily as an inhibitor of reactive oxygen species (ROS) in biochemical and preservation studies. Supplied as a solid, it is intended for laboratory research use only and requires sealed storage away from moisture.

  • Inhibits reactive oxygen species (ROS) generation in biochemical assays.
  • White to yellow solid physical form.
  • Purity 98.0% by analysis.
  • Molecular formula C12H10I2 and molecular weight 408.02 g/mol.
  • Packaged as a 25 g quantity for laboratory use.
  • Store sealed, away from moisture; cold storage recommended for solutions.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-81229 Supplier Medchemexpress LLC Supplier No. HYW25030725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diphenyliodonium iodide is a research-grade reagent used primarily as an inhibitor of reactive oxygen species (ROS) in biochemical and preservation studies. Supplied as a solid, it is intended for laboratory research use only and requires sealed storage away from moisture.

  • Inhibits reactive oxygen species (ROS) generation in biochemical assays.
  • White to yellow solid physical form.
  • Purity 98.0% by analysis.
  • Molecular formula C12H10I2 and molecular weight 408.02 g/mol.
  • Packaged as a 25 g quantity for laboratory use.
  • Store sealed, away from moisture; cold storage recommended for solutions.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.