Learn More
Medchemexpress LLC Diphenyliodonium iodide | 2217-79-0 | MFCD00011910 | 98.0% | 408.02 g/mol | C12H10I2 | 5 G
SDP
Catalog No. 5000281695
Encompass_Preferred
Diphenyliodonium iodide is an organoiodine compound used as a research reagent and inhibitor of reactive oxygen species (ROS). Supplied as a solid powder for laboratory research use only, it is commonly used in biochemical assays to study ROS-related pathways and in photochemical applications.
  • Inhibits reactive oxygen species for ROS modulation studies.
  • Provided as a stable solid powder for easy handling and storage.
  • Suitable for biochemical and cell-based assays.
  • High purity supports reproducible experimental results.
  • Stored refrigerated to maintain stability.
Catalog No. 50-002-81695 Supplier Medchemexpress LLC Supplier No. HYW2503075G
Diphenyliodonium iodide is an organoiodine compound used as a research reagent and inhibitor of reactive oxygen species (ROS). Supplied as a solid powder for laboratory research use only, it is commonly used in biochemical assays to study ROS-related pathways and in photochemical applications.
  • Inhibits reactive oxygen species for ROS modulation studies.
  • Provided as a stable solid powder for easy handling and storage.
  • Suitable for biochemical and cell-based assays.
  • High purity supports reproducible experimental results.
  • Stored refrigerated to maintain stability.