Learn More
AdipoGen Diphenyliodonium nitrate
SDP
Catalog No. 502561281
Encompass_Preferred
Chemical. CAS 722-56-5. Formula C12H10INO3. MW 343.12. Synthetic. Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.
Catalog No. 50-256-1281 Supplier AdipoGen Supplier No. CDXD0046G050
Chemical. CAS 722-56-5. Formula C12H10INO3. MW 343.12. Synthetic. Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.