Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyliodonium Nitrate, TCI America™
SDP

Catalog No. D23575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D23575G Supplier TCI America Supplier No. D23575G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 722-56-5
Molecular Formula C12H10INO3
Molecular Weight (g/mol) 343.12
MDL Number MFCD00031704
InChI Key CQZCVYWWRJDZBO-UHFFFAOYSA-N
Synonym diphenyliodonium nitrate, iodonium, diphenyl-, nitrate, diphenyliodanium nitrate, acmc-1belb
PubChem CID 12876
IUPAC Name diphenyliodanium;nitrate
SMILES C1=CC=C(C=C1)[I+]C2=CC=CC=C2.[N+](=O)([O-])[O-]

Specifications

Color White
Quantity 5 g
UN Number 1479
Formula Weight 343.12
Physical Form Crystalline Powder
Chemical Name or Material Diphenyliodonium Nitrate
EINECSNumber (3)-0030
RTECSNumber NN6665000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.