Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyliodonium Perchlorate 98.0+%, TCI America™
SDP

Catalog No. D22431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22431G Supplier TCI America Supplier No. D22431G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75007-13-5
Molecular Formula C12H10ClIO4
Molecular Weight (g/mol) 380.562
MDL Number MFCD00191355
InChI Key JMJGNDIWCVQJJZ-UHFFFAOYSA-M
PubChem CID 19708536
IUPAC Name diphenyliodanium;perchlorate
SMILES C1=CC=C(C=C1)[I+]C2=CC=CC=C2.[O-]Cl(=O)(=O)=O

Specifications

Melting Point 176°C
Color White
Quantity 1 g
UN Number 1479
Formula Weight 380.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyliodonium Perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.