Learn More
AdipoGen Diphenyliodonium PF6
SDP
Catalog No. 502561284
Encompass_Preferred
Chemical. CAS 58109-40-3. Formula C12H10F6IP. MW 426.08. Synthetic. Catalyst for the photochemical polymerization of various monomers.
Catalog No. 50-256-1284 Supplier AdipoGen Supplier No. CDXD0045G005
Chemical. CAS 58109-40-3. Formula C12H10F6IP. MW 426.08. Synthetic. Catalyst for the photochemical polymerization of various monomers.