Learn More
AdipoGen Diphenyliodonium triflate (5 g)
SDP
Supplier:  AdipoGen CDXD0091G005
Encompass_Preferred
Chemical. CAS: 66003-76-7. Formula: C13H10F3IO3S. MW: 430.18. Synthetic. Reagent for synthesis. Used for alpha-phenylation of cyclic ketones, N-, O-, P-, and S-phenylation reactions. Phenylating agent for palladium-catalyzed cross-couplings, click chemistry and photochemistry.
Catalog No. 50-205-4368