Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyliodonium trifluoromethanesulfonate, 99%

Catalog No. AC432840010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC432840010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC432840010 Supplier Thermo Scientific Chemicals Supplier No. 432840010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 66003-76-7 | C12H10I·CF3O3S | 430.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66003-76-7
Molecular Formula C12H10I·CF3O3S
Molecular Weight (g/mol) 430.18
MDL Number MFCD00191356
InChI Key SBQIJPBUMNWUKN-UHFFFAOYSA-M
Synonym diphenyliodonium trifluoromethanesulfonate, diphenyliodanium triflate, diphenyliodonium triflate, acmc-209nt6, diphenyliodonium trifluoromethansulfonat, diphenyliodanium; trifluoromethanesulfonate, diphenyliodonium trifluoromethane sulfonate, diphenyliodonium; trifluoromethanesulfonate, diphenyliodanium; tris fluoranyl methanesulfonate
PubChem CID 2737137
IUPAC Name diphenyliodanium;trifluoromethanesulfonate
SMILES C1=CC=C(C=C1)[I+]C2=CC=CC=C2.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 179°C to 182°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 430.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Diphenyliodonium trifluoromethanesulfonate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.