Learn More
Medchemexpress LLC Diphenylmethane | 101-81-5 | 99.7% | 168.24 | 100 G
SDP
Catalog No. 5000452197
Encompass_Preferred
Diphenylmethane (Benzylphenyl) is an aromatic compound utilized as a drug intermediate. In the fragrance industry, it functions as an oxidizing agent and is found in soaps. It also serves as a synergist in pesticides and a plasticizer in dyes.
  • Aromatic compound
  • Used as a drug intermediate
  • Used as an oxidizing agent in the fragrance industry and in soaps
  • Acts as a synergist in pesticides
  • Functions as a plasticizer in dyes
Catalog No. 50-004-52197 Supplier Medchemexpress LLC Supplier No. HYW008814100G
Add to Cart
Add to Cart
Diphenylmethane (Benzylphenyl) is an aromatic compound utilized as a drug intermediate. In the fragrance industry, it functions as an oxidizing agent and is found in soaps. It also serves as a synergist in pesticides and a plasticizer in dyes.
  • Aromatic compound
  • Used as a drug intermediate
  • Used as an oxidizing agent in the fragrance industry and in soaps
  • Acts as a synergist in pesticides
  • Functions as a plasticizer in dyes