Learn More
Medchemexpress LLC Diphenylmethane | 101-81-5 | MFCD00004781 | 99.65% | 168.24 | 1 KG
SDP
Catalog No. 5000366310
Encompass_Preferred
Diphenylmethane (Benzylphenyl) is an aromatic compound used as a drug intermediate. In the fragrance industry, it is used as an oxidizing agent and in soaps; in pesticides, as a synergist; and in dyes, as a plasticizer. It is for research use only and not sold to patients.
  • Aromatic compound used as a drug intermediate.
  • Used in the fragrance industry as an oxidizing agent and in soaps.
  • Functions as a synergist in pesticides.
  • Acts as a plasticizer in dyes.
Catalog No. 50-003-66310 Supplier Medchemexpress LLC Supplier No. HYW0088141KG
Add to Cart
Add to Cart
Diphenylmethane (Benzylphenyl) is an aromatic compound used as a drug intermediate. In the fragrance industry, it is used as an oxidizing agent and in soaps; in pesticides, as a synergist; and in dyes, as a plasticizer. It is for research use only and not sold to patients.
  • Aromatic compound used as a drug intermediate.
  • Used in the fragrance industry as an oxidizing agent and in soaps.
  • Functions as a synergist in pesticides.
  • Acts as a plasticizer in dyes.