Learn More
Medchemexpress LLC Diphenylmethanimine | 1013-88-3 | 181.23 | 1000 G
SDP
Catalog No. 5000448791
Encompass_Preferred
Diphenylmethanimine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is useful in the preparation of nitrogen ylide dimers and as an ammonia surrogate. It also serves as an intermediate material for the synthesis of organic chemicals, for example, in the catalytic enantioselective alkylation of glycine t-butyl ester's benzophenone imine.
  • Biochemical reagent
  • Can be used as a biological material or organic compound
  • Useful for life science related research
  • Intermediate material for synthesis of organic chemicals
  • Can be used in catalytic enantioselective alkylation for amino acid derivatives
Catalog No. 50-004-48791 Supplier Medchemexpress LLC Supplier No. HYW0076161000G
Diphenylmethanimine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is useful in the preparation of nitrogen ylide dimers and as an ammonia surrogate. It also serves as an intermediate material for the synthesis of organic chemicals, for example, in the catalytic enantioselective alkylation of glycine t-butyl ester's benzophenone imine.
  • Biochemical reagent
  • Can be used as a biological material or organic compound
  • Useful for life science related research
  • Intermediate material for synthesis of organic chemicals
  • Can be used in catalytic enantioselective alkylation for amino acid derivatives