Learn More
Medchemexpress LLC Diphenylmethanimine | 1013-88-3 | 181.23 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW007616500G
Encompass_Preferred
Diphenylmethanimine is a biochemical reagent useful in life science related research. It serves as a biological material or organic compound, applicable in the preparation of nitrogen ylide dimers, and as an ammonia surrogate, also functioning as an intermediate for organic chemical synthesis.
  • Useful in the preparation of nitrogen ylide dimers.
  • Functions as a benzophenone imine as ammonia surrogate.
  • An intermediate material for the synthesis of organic chemicals.
  • Catalytic enantioselective alkylation of the benzophenone imine of glycine t-butyl ester using phosphazene (Schwesinger) bases and chiral quaternary ammonium salts yields either enantiomer of amino acid derivatives in excellent yield.
Catalog No. 50-004-48737