Learn More
Medchemexpress LLC Diphenylmethanol-d5 | 95450-78-5 | 189.26 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW004059S10MG
Encompass_Preferred
Diphenylmethanol-d5 is the deuterium labeled Diphenylmethanol.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements, like deuterium, have gained attention for their potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-47143