Learn More
Medchemexpress LLC Diphenylmethanol-d5 | 95450-78-5 | 189.26 | 25 MG
SDP
Catalog No. 5000449388
Encompass_Preferred
Diphenylmethanol-d5 is a deuterium labeled compound primarily used in drug development. Incorporating stable heavy isotopes like deuterium allows its use as a tracer for quantitation and has the potential to impact drug pharmacokinetics and metabolic profiles.
  • Deuterium labeled Diphenylmethanol
  • Used as a tracer for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Can be used as an internal standard for quantitative analysis (e.g., by NMR, GC-MS, or LC-MS)
  • High purity of 99%
  • Molecular weight of 189.26
Catalog No. 50-004-49388 Supplier Medchemexpress LLC Supplier No. 25MGHYW004059S
Diphenylmethanol-d5 is a deuterium labeled compound primarily used in drug development. Incorporating stable heavy isotopes like deuterium allows its use as a tracer for quantitation and has the potential to impact drug pharmacokinetics and metabolic profiles.
  • Deuterium labeled Diphenylmethanol
  • Used as a tracer for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Can be used as an internal standard for quantitative analysis (e.g., by NMR, GC-MS, or LC-MS)
  • High purity of 99%
  • Molecular weight of 189.26