Learn More
Medchemexpress LLC Diphenylmethanol-d5 | 95450-78-5 | 189.26 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW004059S5MG
Encompass_Preferred
Diphenylmethanol-d5 is a deuterium labeled Diphenylmethanol, for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitative analysis
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-47146