Learn More
Medchemexpress LLC Diphenylmethanol-d5 | 95450-78-5 | 189.26 | 50 MG
SDP
Catalog No. 5000445873
Encompass_Preferred
Diphenylmethanol-d5 is a deuterium labeled form of Diphenylmethanol. It is suitable for use as a tracer or as an internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS. Deuteration of drug molecules has shown potential to influence their pharmacokinetic and metabolic profiles.
  • Deuterium labeled Diphenylmethanol
  • Suitable as a tracer
  • Internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-45873 Supplier Medchemexpress LLC Supplier No. HYW004059S50MG
Diphenylmethanol-d5 is a deuterium labeled form of Diphenylmethanol. It is suitable for use as a tracer or as an internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS. Deuteration of drug molecules has shown potential to influence their pharmacokinetic and metabolic profiles.
  • Deuterium labeled Diphenylmethanol
  • Suitable as a tracer
  • Internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Potential to affect pharmacokinetic and metabolic profiles of drugs