Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylmethyl 7beta-Amino-3-cephem-4-carboxylate 98.0+%, TCI America™
SDP

Catalog No. D52341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D52341G Supplier TCI America Supplier No. D52341G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36923-21-4
Molecular Formula C20H18N2O3S
Molecular Weight (g/mol) 366.44
MDL Number MFCD23701579
InChI Key NYYBPASOTOAXQW-UHFFFAOYNA-N
Synonym Benzhydryl (6R,7R)-7-Amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, (6R,7R)-7-Amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Benzhydryl Ester, 7beta-Amino-3-cephem-4-carboxylic Acid Diphenylmethyl Ester
PubChem CID 93477338
IUPAC Name diphenylmethyl 7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES NC1C2SCC=C(N2C1=O)C(=O)OC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 166°C
Color Yellow
Quantity 1 g
Formula Weight 366.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylmethyl 7beta-Amino-3-cephem-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.