Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diphenylphosphinic Chloride

Catalog No. 23023550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-023-550G 50 g
23-023-510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 23-023-550G Supplier Sigma Aldrich Supplier No. 23023550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diphenylphosphinic chloride (Ph2POCl) is widely used as a precursor to synthesize various chiral phosphine bidentate ligands for asymmetric synthesis, in addition to several peptide coupling agents.

Specifications

CAS 1499-21-4
Density 1.24 g/mL at 25°C
Boiling Point 222°C (16 mmHg, lit.)
Molecular Formula C12H10ClOP
Linear Formula (C6H5)2P(O)Cl
Refractive Index n20/D 1.609
MDL Number MFCD00002077
Quantity 50 g
Synonym Diphenylchlorophosphine oxide; Diphenylphosphinic acid chloride; Diphenylphosphinoyl chloride; Diphenylphosphinyl chloride; Diphenylphosphorus oxychloride; Ph2POCl; Chlorodiphenylphosphine oxide
Molecular Weight (g/mol) 236.63 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.