Learn More
Cayman Chemical Diphenylpyraline, Purity: ≥95%, CAS: 147-20-6, 250 mg
SDP
Catalog No. 502949460
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 20 nM); inhibits ovalbumin-induced bronchoconstriction in ovalbumin-sensitized guinea pigs at 20 µmol/kg; reduces dopamine uptake in rat nucleus accumbens at 14 mg/kg
Catalog No. 50-294-9460 Supplier Cayman Chemical Supplier No. 33237250 MG
A histamine H1 receptor antagonist (Ki = 20 nM); inhibits ovalbumin-induced bronchoconstriction in ovalbumin-sensitized guinea pigs at 20 µmol/kg; reduces dopamine uptake in rat nucleus accumbens at 14 mg/kg