Learn More
Apexbio Technology LLC Diphenylpyraline HCl 132-18-3 1g
SDP
Catalog No. 5000093856
Encompass_Preferred
Diphenylpyraline HCl (CAS 132-18-3) is a small molecule classified within the diphenylpiperidine group and functions as a first-generation antihistamine It acts by competitively inhibiting histamine H1 receptors thereby attenuating histamine-mediated cellular responses In addition to its antihistaminic activity diphenylpyraline exhibits anticholinergic properties influencing neurotransmitter regulation within the central and peripheral nervous systems This compound is widely utilized in the study of histaminergic and cholinergic pathways as well as for elucidating mechanisms underlying allergic reactions and central nervous system disorders
Catalog No. 50-000-93856 Supplier Apexbio Technology LLC Supplier No. A84151000
Add to Cart
Add to Cart
Diphenylpyraline HCl (CAS 132-18-3) is a small molecule classified within the diphenylpiperidine group and functions as a first-generation antihistamine It acts by competitively inhibiting histamine H1 receptors thereby attenuating histamine-mediated cellular responses In addition to its antihistaminic activity diphenylpyraline exhibits anticholinergic properties influencing neurotransmitter regulation within the central and peripheral nervous systems This compound is widely utilized in the study of histaminergic and cholinergic pathways as well as for elucidating mechanisms underlying allergic reactions and central nervous system disorders