Learn More
Medchemexpress LLC Diphenylpyraline hydrochloride | 132-18-3 | 99.9% | 1 ML
SDP
Catalog No. 5000407983
Encompass_Preferred
Diphenylpyraline hydrochloride is a potent histamine H1 receptor antagonist, functioning as an orally active antihistamine agent with antimuscarinic and antiallergic effects.
  • Relieves allergic conditions such as rhinitis and hay fever.
  • Addresses pruritic skin disorders in vivo.
  • Inhibits dopamine uptake in mouse nucleus accumbens slices.
  • Elevates extracellular dopamine levels in mouse nucleus accumbens.
  • Induces locomotor activation in mice.
Catalog No. 50-004-07983 Supplier Medchemexpress LLC Supplier No. HYB097010MM/1ML
Diphenylpyraline hydrochloride is a potent histamine H1 receptor antagonist, functioning as an orally active antihistamine agent with antimuscarinic and antiallergic effects.
  • Relieves allergic conditions such as rhinitis and hay fever.
  • Addresses pruritic skin disorders in vivo.
  • Inhibits dopamine uptake in mouse nucleus accumbens slices.
  • Elevates extracellular dopamine levels in mouse nucleus accumbens.
  • Induces locomotor activation in mice.