Learn More
Medchemexpress LLC Diphenylpyraline (hydrochloride) | 132-18-3 | 99.9% | 500 MG
SDP
Catalog No. 5000407628
Encompass_Preferred
Diphenylpyraline hydrochloride is a potent histamine H1 receptor antagonist and an orally active antihistamine agent. It exhibits antimuscarinic and antiallergic effects, making it suitable for the relief of allergic conditions such as rhinitis, hay fever, and pruritic skin disorders.
  • Potent histamine H1 receptor antagonist
  • Orally active antihistamine agent
  • Exhibits antimuscarinic effects
  • Provides antiallergic effects
  • Suitable for rhinitis and hay fever relief
  • Used for pruritic skin disorders
  • Inhibits dopamine uptake in mouse nucleus accumbens slices
  • Elevates extracellular dopamine levels and induces locomotor activation in mice
Catalog No. 50-004-07628 Supplier Medchemexpress LLC Supplier No. HYB0970500MG
Diphenylpyraline hydrochloride is a potent histamine H1 receptor antagonist and an orally active antihistamine agent. It exhibits antimuscarinic and antiallergic effects, making it suitable for the relief of allergic conditions such as rhinitis, hay fever, and pruritic skin disorders.
  • Potent histamine H1 receptor antagonist
  • Orally active antihistamine agent
  • Exhibits antimuscarinic effects
  • Provides antiallergic effects
  • Suitable for rhinitis and hay fever relief
  • Used for pruritic skin disorders
  • Inhibits dopamine uptake in mouse nucleus accumbens slices
  • Elevates extracellular dopamine levels and induces locomotor activation in mice