Learn More
Strem, An Ascensus Company CAS# 775-12-2. 5g. Diphenylsilane, min. 97%. MFCD00003002
SDP
Catalog No. 501385555
Encompass
CAS# 775-12-2. 5g. Diphenylsilane, min. 97%. MFCD00003002. Molecular Weight: 184.30. Molecular Formula: (C6H5)2SiH2. Color/form: colorless liq. Strem# 93-1485. http://www.strem.com/catalog/v/93-1485/
Catalog No. 50-138-5555 Supplier Strem, An Ascensus Company Supplier No. 9314855G
CAS# 775-12-2. 5g. Diphenylsilane, min. 97%. MFCD00003002. Molecular Weight: 184.30. Molecular Formula: (C6H5)2SiH2. Color/form: colorless liq. Strem# 93-1485. http://www.strem.com/catalog/v/93-1485/