Learn More
Medchemexpress LLC Diphenylsulfane-d1 | 180802-01-1 | 99.5% | 187.28 g/mol | C12H9DS | 5 MG
SDP
Catalog No. 5000302153
Encompass_Preferred
Diphenylsulfane-d1 is the deuterium-labeled analogue of diphenylsulfane (diphenyl sulfide), supplied in small research quantities as an isotopically labeled reference or tracer for analytical, mechanistic, and mass spectrometry applications.
  • Deuterium-labeled at one position (d1) for isotopic tracing.
  • High chemical purity suitable for use as an analytical standard.
  • Provided in small-quantity packaging tailored for research use.
  • Suitable as an internal standard in mass spectrometry and quantitative analysis.
  • Chemical identity corresponds to formula C12H9DS and molar mass ~187.28 g/mol.
Catalog No. 50-003-02153 Supplier Medchemexpress LLC Supplier No. HYW016174S5MG
Diphenylsulfane-d1 is the deuterium-labeled analogue of diphenylsulfane (diphenyl sulfide), supplied in small research quantities as an isotopically labeled reference or tracer for analytical, mechanistic, and mass spectrometry applications.
  • Deuterium-labeled at one position (d1) for isotopic tracing.
  • High chemical purity suitable for use as an analytical standard.
  • Provided in small-quantity packaging tailored for research use.
  • Suitable as an internal standard in mass spectrometry and quantitative analysis.
  • Chemical identity corresponds to formula C12H9DS and molar mass ~187.28 g/mol.