Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyltin oxide, Thermo Scientific Chemicals

Catalog No. AA3961514
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3961514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3961514 Supplier Thermo Scientific Chemicals Supplier No. 03961514
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2273-51-0 | Molecular Formula: C12H10OSn | Molecular Weight: 288.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2273-51-0
Molecular Formula C12H10OSn
Molecular Weight (g/mol) 288.92
MDL Number MFCD00058851
InChI Key VPPWQRIBARKZNY-UHFFFAOYSA-N
Synonym diphenyltin oxide, diphenyltin oxide polymer, stannane, diphenyloxo-, polymer, stannane, oxodiphenyl-, polymer, diphenylstannanone, oxodiphenylstannane, oxo diphenyl stannane, oxo diphenyl tin, stannane, polymer, iv
PubChem CID 75280
IUPAC Name oxo(diphenyl)tin
SMILES O=[Sn](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 320°C (decomposition)
Quantity 25 g
UN Number UN3146
Formula Weight 288.9
Physical Form Powder
Chemical Name or Material Diphenyltin oxide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 218-883-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.