Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyltin sulfide, 99%, Thermo Scientific™

Catalog No. AA3961403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3961403 1 g
AA3961406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3961403 Supplier Thermo Scientific Chemicals Supplier No. 3961403

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20332-10-9
Molecular Formula C12H10SSn
Molecular Weight (g/mol) 304.982
MDL Number MFCD00058856
InChI Key CYXRBVIJBDYNQU-UHFFFAOYSA-N
Synonym diphenyltin sulfide, diphenylthioxostannane, diphenylstannanethione, diphenyl tin sulfide, diphenyl thioxo stannane, ph2sns, acmc-209ums, diphenyl sulfanylidene tin
PubChem CID 88492
IUPAC Name diphenyl(sulfanylidene)tin
SMILES C1=CC=C(C=C1)[Sn](=S)C2=CC=CC=C2

Specifications

Quantity 1 g
UN Number UN3146
Formula Weight 304.97
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Diphenyltin sulfide
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 243-739-6
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.