Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyltin Sulfide 97.0+%, TCI America™
SDP

Catalog No. D23581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D23581G Supplier TCI America Supplier No. D23581G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20332-10-9
Molecular Formula C12H10SSn
Molecular Weight (g/mol) 304.982
MDL Number MFCD00058856
InChI Key CYXRBVIJBDYNQU-UHFFFAOYSA-N
Synonym diphenyltin sulfide, diphenylthioxostannane, diphenylstannanethione, diphenyl tin sulfide, diphenyl thioxo stannane, ph2sns, acmc-209ums, diphenyl sulfanylidene tin
PubChem CID 88492
IUPAC Name diphenyl(sulfanylidene)tin
SMILES C1=CC=C(C=C1)[Sn](=S)C2=CC=CC=C2

Specifications

Melting Point 185°C
Color White
Quantity 1 g
UN Number 3146
Formula Weight 304.98
Percent Purity ≥97.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyltin Sulfide [Activator for O-Glycoside Synthesis]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.