Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diphosphoryl Chloride

Catalog No. 38182925ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-182-925ML 25 mL
38-182-95ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 38-182-925ML Supplier Sigma Aldrich Supplier No. 38182925ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Useful reagent for the Vilsmeier formylation and glyoxylation reactions of nucleophilic aromatic compounds.

Specifications

CAS 13498-14-1
Density 1.82 g/mL at 25°C
Boiling Point 90°C (12 mmHg, lit.)
Molecular Formula Cl4O3P2
Refractive Index n20/D 1.475
Linear Formula Cl2P(O)OP(O)Cl2
Quantity 25 mL
MDL Number MFCD00061542
Synonym Diphosphoric tetrachloride; Pyrophosphoric tetrachloride; Pyrophosphoryl chloride
Molecular Weight (g/mol) 251.76 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.