Learn More
As One International Inc Diphtheria toxin mutant CRM197, active but nontoxic, 200ug
Catalog No. 501654923
Encompass_Preferred
The CRM197 was highly purified (>95% pure) from cultured medium of Corynebacterium diphtheriae mutant CRM197. CRM197 can bind to HB-EGF and inhibit the growth stimulating activity of HB-EGF without showing cellular toxicity.Form: 1.0 mg/ ml in 20 mM Tris-HCl (pH 7.2) 150 mM NaClShip with dry-ice and store at -80℃Applications: 1. Inhibitor of HB-EGF2. Candidate of anticancer drug for tumors whose growth depends on HB-EGF
Catalog No. 50-165-4923 Supplier As One International Inc Supplier No. 01515
The CRM197 was highly purified (>95% pure) from cultured medium of Corynebacterium diphtheriae mutant CRM197. CRM197 can bind to HB-EGF and inhibit the growth stimulating activity of HB-EGF without showing cellular toxicity.Form: 1.0 mg/ ml in 20 mM Tris-HCl (pH 7.2) 150 mM NaClShip with dry-ice and store at -80℃Applications: 1. Inhibitor of HB-EGF2. Candidate of anticancer drug for tumors whose growth depends on HB-EGF